VWR® Graefe Micro Hook
VWR® Graefe Micro Hook, Steel, 11.4 cm (4.5"), Sharp
Catalog # 10806-340
Supplier: VWR International
Jump to:
Product Details & Documents
The VWR® Graefe Micro Hook is constructed from German steel.
- Passivated
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Devices are available in sharp or blunt formats.
Spécifications
- Désignation:VWR® Graefe Micro Hook
- Longueur:11.4 cm (4.5")
- Matière:Steel
- Tip Style:Sharp
- Cat. No.:10806-340