Interlocking Storage Cabinets, Quantum Storage Systems
Interlocking cabinet, 10 drawers, 11¾", 11⅜", 11"
10808-768
: QIC-4163GY
Interlocking storage cabinets have the unique ability to interlock horizontally and vertically, allowing customization based on storage needs.
- Pre-drilled holes on the backside of the cabinet allow for wall mounting
- Cabinets are made from high impact polystyrene
- Drawers are removable and will not rust or corrode
- Drawers have a recessed handle and come with a slotted label holder
Drawers have seven divider slots molded in so the drawer can be divided into multiple compartments. Clear window allows for easy identification of parts. Nine pre-configured storage system comes with drawers, clear windows and two dividers per drawer.
Drawers 11 x2 3/4 x2 1/2" Or 11 x5 5/8 x 2 1/2"
: Additional dividers can be purchased separately.
- Profondeur:11⅜"
- Largeur:11¾"
- Désignation:Interlocking cabinet
- Hauteur:11"
- Type:10 drawers