Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
pH Buffer Solutions, NIST

pH Buffer Solutions, NIST

Bottle, 500 ml, 8.00
Catalog # 10804-858
Supplier:   Mettler Toledo
Supplier #:  30258827
pH Buffer Solutions, NIST
Catalog # 10804-858
Supplier:  Mettler Toledo
CAS Number:  
pH Buffer Solutions, NIST
pH Buffer Solutions, NIST

Bottle, 500 ml, 8.00

CAS Number:  
Price
$53.20
Log in pour la disponibilité
Each (500ml)
Product Details & Documents

These buffer solutions are used for calibration and verification of measurement system.

  • Shelf life: 24 months
  • Uncertainty: ±0.02 pH

Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Spécifications

Spécifications

  • Type de conditionnement:
    Bottle
  • Taille:
    500 ml
  • pH:
    8.00

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.