Screw Caps, MicroSolv™
PP, White Silicone Rubber / White PTFE Pre-Slit, fitted, 9-425 mm, Blue, Screw cap
Catalog # 10804-658
Supplier: MicroSolv Technology Corp
Fournisseur #: 9502S-30M-B
Jump to:
Product Details & Documents
Spécifications
- Taille du bouchon:9-425 mm
- Cap Type:Screw cap
- Septum:White Silicone Rubber / White PTFE Pre-Slit, fitted
- Matière:PP
- Cap Color:Blue