Screw Caps, MicroSolv™
PP, Tan Silicone Rubber / White PTFE, bonded, 9-425 mm, Green, Screw cap
10804-646
: 9502S-11-MG
- Taille du bouchon:9-425 mm
- Cap Type:Screw cap
- Septum:Tan Silicone Rubber / White PTFE, bonded
- Matière:PP
- Cap Color:Green