$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
18.5" X 17-1/8" X 11-1/8" WIRE MESH BIN
10804-548
:
QUANTUM STORAGE SYSTEMS
18.5" X 17-1/8" X 11-1/8" WIRE MESH BIN
10804-548
:
QUANTUM STORAGE SYSTEMS
:
QMB570C
Frequently Bought Together
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH