ESD Modular Trays, Quantum Storage Systems
ESD Trays; Conductive, 20 × 26 × 1 1/2"
Catalog # 10804-488
 :  2026ESD
ESD Modular Trays, Quantum Storage Systems
Catalog # 10804-488
ESD Modular Trays, Quantum Storage Systems
ESD Modular Trays, Quantum Storage Systems
ESD Trays; Conductive, 20 × 26 × 1 1/2"
Price
$357.06
Log in for availability
Each
Product Details & Documents
These trays are ESD and comply with ANSI/ESD standards for the use in PC board assembly and handling.
  • Withstand temperatures of −40 °F to +350 °F
  • Trays are resistant to abrasion and chemicals
  • Trays are permanently conductive, properties are unaffected by washing in steam or water
All trays are manufactured of fiberglass reinforced thermoset material, allowing up to 200 lb. weight capacity when evenly distributed.
Specifications
  • Désignation:
    ESD Trays; Conductive
  • L×W×H:
    20 × 26 × 1 1/2"