Screw Caps, MicroSolv™
PP, Silicone Rubber / PTFE Pre-Slit, 13 - 425 mm, Black, Screw cap
Catalog # 10804-136
Supplier: MicroSolv Technology Corp
Fournisseur #: 9504S-35C-BLK
Jump to:
Product Details & Documents
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
Spécifications
- Taille du bouchon:13 - 425 mm
- Cap Type:Screw cap
- Septum:Silicone Rubber / PTFE Pre-Slit
- Matière:PP
- Cap Color:Black