Screw Caps, MicroSolv™ | Avantor
Screw Caps, MicroSolv™
PP, Silicone Rubber / PTFE Pre-Slit, 13 - 425 mm, Black, Screw cap
Catalog # 10804-136
Fournisseur #:  9504S-35C-BLK
Screw Caps, MicroSolv™
Catalog # 10804-136
Screw Caps, MicroSolv™
Screw Caps, MicroSolv™
PP, Silicone Rubber / PTFE Pre-Slit, 13 - 425 mm, Black, Screw cap
Price
$66.87
Log in pour la disponibilité
Pack of 100
Product Details & Documents
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
Spécifications
  • Taille du bouchon:
    13 - 425 mm
  • Cap Type:
    Screw cap
  • Septum:
    Silicone Rubber / PTFE Pre-Slit
  • Matière:
    PP
  • Cap Color:
    Black