Screw Caps, MicroSolv™
PP, Silicone Rubber / PTFE Pre-Slit, 13 - 425 mm, White, Screw cap
Catalog # 10804-134
Supplier: MicroSolv Technology Corp
Fournisseur #: 9504S-30C-W-K
Jump to:
Product Details & Documents
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
Spécifications
- Taille du bouchon:13 - 425 mm
- Cap Type:Screw cap
- Septum:Silicone Rubber / PTFE Pre-Slit
- Matière:PP
- Cap Color:White