Low-Halide Polyester Labels, B30, Brady | Avantor
Low-Halide Polyester Labels, B30, Brady
Catalog # 10018-486
Low-Halide Polyester Labels, B30, Brady
Catalog # 10018-486
Supplier:  
Low-Halide Polyester Labels, B30, Brady
Low-Halide Polyester Labels, B30, Brady
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
B30 B569 LT BLU 4INX100FT Catalog #10018-462
Product Details & Documents
Hi-Performance Polyester Labels (B-569) have a low halide content and are designed for marking stainless steel pipes, particularly in harsh environments.
  • Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
  • Outdoor grade polyester label can be thermal transfer printed
  • Withstands mild chemicals, oil, and water
  • Service temperature of –40 to 100 °C (–40 to 212 °F)
  • For use with BBP®30; BBP®31; BBP®33; BBP®35; BBP®37 printers
Applies best to very smooth surfaces and ideal for mounting to rigid panels. Continuous roll is not cut to length. It is cut by the printer.