Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Low-Halide Polyester Labels, B30, Brady

Low-Halide Polyester Labels, B30, Brady

Low-halide polyester labels, B-569, Polyester, Pink, 1.125", 30.48 m (100 ft.)
  10018-000
Supplier #:  117967
Low-Halide Polyester Labels, B30, Brady
  10018-000
Low-Halide Polyester Labels, B30, Brady
Low-Halide Polyester Labels, B30, Brady

Low-halide polyester labels, B-569, Polyester, Pink, 1.125", 30.48 m (100 ft.)

 
$408.34
 
   
Each
 

Hi-Performance Polyester Labels (B-569) have a low halide content and are designed for marking stainless steel pipes, particularly in harsh environments.

  • Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
  • Outdoor grade polyester label can be thermal transfer printed
  • Withstands mild chemicals, oil, and water
  • Service temperature of –40 to 100 °C (–40 to 212 °F)
  • For use with BBP®30; BBP®31; BBP®33; BBP®35; BBP®37 printers

Applies best to very smooth surfaces and ideal for mounting to rigid panels. Continuous roll is not cut to length. It is cut by the printer.

 
  • Color:
    Pink
  • Désignation:
    Low-halide polyester labels, B-569
  • Matière:
    Polyester
  • Roll Length:
    30.48 m (100 ft.)
  • Width:
    1.125"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.