Ward's® Basic Electrophoresis Bundles | Avantor
Ward's® Basic Electrophoresis Bundles
Large-Basic Bundle, 6 Gel Boxes; 3 Power Supply; 12 Fixed-Volume Pipettes with Tips; 3 Kits with Dyes
Catalog # 470218-344
Supplier:   Avantor
Ward's® Basic Electrophoresis Bundles
Catalog # 470218-344
Supplier:  Avantor
Ward's® Basic Electrophoresis Bundles
Ward's® Basic Electrophoresis Bundles
Large-Basic Bundle, 6 Gel Boxes; 3 Power Supply; 12 Fixed-Volume Pipettes with Tips; 3 Kits with Dyes
Price
$2,944.75
Log in pour la disponibilité
Each Kit of 1
Product Details & Documents
These bundles are everything you need to perform agarose gel electrophoresis.
  • Each bundle contains everything you need to get started with Electrophoresis in your lab
  • Included kit(s) come with easy-to-use electrophoretic dyes and detailed instructions
  • Choose your bundle size based on the size of your class
Gel electrophoresis is a great way to introduce your students to biotechnology and reinforce key genetics concepts.
These convenient bundles are designed to provide you with all of the materials you need to perform gel electrophoresis with your students. Choose the size of equipment bundle that you need based on your class size. Our basic systems offer an affordable solution with easy-to-use electrophoretic dyes.
Ordering information: Small Basic Bundle contains 2 gel boxes; 1 power supply; 4 fixed-volume (10μL) micropipettes; 2 racks of tips; 1 kit: "Introduction to Gel Electrophoresis with Dyes.
Medium Basic Bundle contains 4 gel boxes; 2 power supplies; 8 fixed-volume (10μL) micropipettes; 4 racks of tips; 2 kits: "Introduction to Gel Electrophoresis with Dyes.
Large Basic Bundle contains 6 gel boxes; 3 power supplies; 12 fixed-volume (10μL) micropipettes; 10 racks of tips; 3 kits: "Introduction to Gel Electrophoresis with Dyes.
Spécifications
  • Désignation:
    Large-Basic Bundle
  • Includes:
    6 Gel Boxes; 3 Power Supply; 12 Fixed-Volume Pipettes with Tips; 3 Kits with Dyes