Glassware Washers Accessories | Avantor
Glassware Washers Accessories
Basket and cover, stainless steel, general use (small glassware), 7.5L×7.5Wx4"D, ¼ of rack, top or bottom
Catalog # 10002-904
Supplier:   VWR International
Glassware Washers Accessories
Catalog # 10002-904
Supplier:  VWR International
Glassware Washers Accessories
Glassware Washers Accessories
Basket and cover, stainless steel, general use (small glassware), 7.5L×7.5Wx4"D, ¼ of rack, top or bottom
Price
$672.79
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Accessories designed for use with VWR® Undercounter, Freestanding and Vertical Stacking Glassware washers safeguard flasks, beakers, test tubes, bottles, Petri dishes, and more.
  • Accessories maximize space, improve efficiency, and protect specialty glassware; use in standard open racks
  • Bottom mesh inserts prevent small items from falling through
  • Made of stainless steel for long life and durability
  • Booster pump for use with low pressure DI water supply below 18 psi
See details in table below for specific use information. For further assistance, view the product literature or contact your Avantor representative.
Spécifications
  • Désignation:
    Basket and cover, stainless steel, general use (small glassware), 7.5L×7.5Wx4"D
  • Fits:
    ¼ of rack, top or bottom
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.