U-2D™ Micro-sample management system with racked inserts
Product Details & Documents
Manage multiple low volume samples in a 96 well format and a unique split rack with glass inserts for vials.
- 350 µl inserts can be used individually by dropping them into a 2 ml vial
- Compatible with plate readers
- Fill the inserts with multi-channel pipettors for quick and easy sample preparation
- Transport inserts safely
Use this unique sample management system that is designed for preparing samples or for injecting them with autosamplers. Each rack is supplied with 96 glass inserts that are installed and ready for use. The included insert rack can be split from the base allowing full visual inspection of the insert bottoms to identify and remove bubble formation. Achieve even and thorough thermal control of all samples by inserting into a water bath or using other heat or cooling methods. The complete rack and inserts are compatible with all autosamplers that accept the ANSI standard 96-well format if desired.
Spécifications
- Désignation:Preloaded U-2D™ plate system rack. Includes 96, clear, 350 µl glass low volume precision inserts with wide conical points
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Zone Free Sealing Film for U-2D™ Sealing FilmZone Free Sealing Film for U-2D™ Sealing Film10002-828Log in pour la disponibilitéCase of 50 - $75.97
Informations sur le produit
- Catalog #AvailabilityPrice
- Permanent Black Marker, for use with U-2D™ Plate SystemPermanent Black Marker, for use with U-2D™ Plate System10002-812Log in pour la disponibilitéEach - $8.01
Informations sur le produit
- Catalog #AvailabilityPrice
- Permanent Red Marker, for use with U-2D™ Plate SystemPermanent Red Marker, for use with U-2D™ Plate System10002-816Log in pour la disponibilitéEach - $7.62
Informations sur le produit
- Catalog #AvailabilityPrice
- U-2D™ plate system easy purchase pack with rack and base only. Includes 96, clear, 350 µl glass low volume precision inserts with wide conical pointsU-2D™ plate system easy purchase pack with rack and base only. Includes 96, clear, 350 µl glass low volume precision inserts with wide conical points10014-946Log in pour la disponibilitéEach - $201.25
Informations sur le produit
- Catalog #AvailabilityPrice
- Permanent Red Marker, for use with U-2D™ Plate SystemPermanent Red Marker, for use with U-2D™ Plate System10002-818Log in pour la disponibilitéPack of 10 - $57.86
Informations sur le produit
- Catalog #AvailabilityPrice
- Permanent Silver Marker, for use with U-2D™ Plate System, Extra Fine TipPermanent Silver Marker, for use with U-2D™ Plate System, Extra Fine Tip75812-810Log in pour la disponibilitéEach - $20.54
Informations sur le produit
- Catalog #AvailabilityPrice
- Permanent Black Marker, for use with U-2D™ Plate System, 0.45mm Fine TipPermanent Black Marker, for use with U-2D™ Plate System, 0.45mm Fine Tip75812-800Log in pour la disponibilitéEach - $19.67
Informations sur le produit
- Catalog #AvailabilityPrice
- Paddles for U-2D™ Sealing Film
Informations sur le produit
- Catalog #AvailabilityPrice
- Permanent Black Marker, for use with U-2D™ Plate SystemPermanent Black Marker, for use with U-2D™ Plate System10002-814Log in pour la disponibilitéPack of 10 - $57.86
Informations sur le produit
- Catalog #AvailabilityPrice
- Roller for U-2D™ Sealing Film
Informations sur le produit
- Catalog #AvailabilityPrice
- Permanent White Marker, for use with U-2D™ Plate System, Extra Fine TipPermanent White Marker, for use with U-2D™ Plate System, Extra Fine Tip75812-812Log in pour la disponibilitéEach - $20.54