U-2D™ Micro-sample management system with racked inserts
Preloaded U-2D™ plate system rack. Includes 96, clear, 350 µl glass low volume deactivated precision inserts with wide conical points
10002-808
: 95296-350-D-PK
Manage multiple low volume samples in a 96 well format and a unique split rack with glass inserts for vials.
- 350 µl inserts can be used individually by dropping them into a 2 ml vial
- Compatible with plate readers
- Fill the inserts with multi-channel pipettors for quick and easy sample preparation
- Transport inserts safely
Use this unique sample management system that is designed for preparing samples or for injecting them with autosamplers. Each rack is supplied with 96 glass inserts that are installed and ready for use. The included insert rack can be split from the base allowing full visual inspection of the insert bottoms to identify and remove bubble formation. Achieve even and thorough thermal control of all samples by inserting into a water bath or using other heat or cooling methods. The complete rack and inserts are compatible with all autosamplers that accept the ANSI standard 96-well format if desired.
- Désignation:Preloaded U-2D™ plate system rack. Includes 96, clear, 350 µl glass low volume deactivated precision inserts with wide conical points
Accessories
- Description
- Description
- Description
- Description
- 10014-946U-2D™ plate system easy purchase pack with rack and base only. Includes 96, clear, 350 µl glass low volume precision inserts with wide conical pointsEach - $201.25
- Description
- Description
- Description
- Description
- Description
- Description
- Description