U-2D™ Micro-sample management system with racked inserts
U-2D™ Micro-Sample Management System. This set includes 96, 350 µl precision, deactivated glass inserts in a rack with a base and a silicone rubber, plug style sealing mat
  10002-796
 :  95296-350-1DP
U-2D™ Micro-sample management system with racked inserts
  10002-796
U-2D™ Micro-sample management system with racked inserts
U-2D™ Micro-sample management system with racked inserts
U-2D™ Micro-Sample Management System. This set includes 96, 350 µl precision, deactivated glass inserts in a rack with a base and a silicone rubber, plug style sealing mat
$259.63
Pack of 1
Manage multiple low volume samples in a 96 well format and a unique split rack with glass inserts for vials.
  • 350 µl inserts can be used individually by dropping them into a 2 ml vial
  • Compatible with plate readers
  • Fill the inserts with multi-channel pipettors for quick and easy sample preparation
  • Transport inserts safely
Use this unique sample management system that is designed for preparing samples or for injecting them with autosamplers. Each rack is supplied with 96 glass inserts that are installed and ready for use. The included insert rack can be split from the base allowing full visual inspection of the insert bottoms to identify and remove bubble formation. Achieve even and thorough thermal control of all samples by inserting into a water bath or using other heat or cooling methods. The complete rack and inserts are compatible with all autosamplers that accept the ANSI standard 96-well format if desired.
  • Désignation:
    U-2D™ Micro-Sample Management System. This set includes 96, 350 µl precision, deactivated glass inserts in a rack with a base and a silicone rubber, plug style sealing mat
Accessories
  •  
    Description
  • 10002-828
    Zone Free Sealing Film for U-2D™ Sealing Film
    Case of 50 - $75.97
  •  
    Description
  • 10002-812
    Permanent Black Marker, for use with U-2D™ Plate System
    Each - $8.01
  •  
    Description
  • 10002-816
    Permanent Red Marker, for use with U-2D™ Plate System
    Each - $7.62
  •  
    Description
  • 10014-946
    U-2D™ plate system easy purchase pack with rack and base only. Includes 96, clear, 350 µl glass low volume precision inserts with wide conical points
    Each - $201.25
  •  
    Description
  • 10002-818
    Permanent Red Marker, for use with U-2D™ Plate System
    Pack of 10 - $57.86
  •  
    Description
  • 75812-810
    Permanent Silver Marker, for use with U-2D™ Plate System, Extra Fine Tip
    Each - $20.54
  •  
    Description
  • 75812-800
    Permanent Black Marker, for use with U-2D™ Plate System, 0.45mm Fine Tip
    Each - $19.67
  •  
    Description
  • 10002-820
    Paddles for U-2D™ Sealing Film
    Pack of 5 - $32.67
  •  
    Description
  • 10002-814
    Permanent Black Marker, for use with U-2D™ Plate System
    Pack of 10 - $57.86
  •  
    Description
  • 10002-822
    Roller for U-2D™ Sealing Film
    Each - $113.36
  •  
    Description
  • 75812-812
    Permanent White Marker, for use with U-2D™ Plate System, Extra Fine Tip
    Each - $20.54