Oxoid CampyGen Compact | Avantor
Oxoid CampyGen Compact
CampyGen compact, 20 sachets
Catalog # CA10790-378
Fournisseur #:  CN020C
Oxoid CampyGen Compact
Catalog # CA10790-378
Oxoid CampyGen Compact
Oxoid CampyGen Compact
CampyGen compact, 20 sachets
Price
$269.00
Log in pour la disponibilité
Each
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
The Oxoid CampyGen and CampyGen Compact are simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.
Like the AnaeroGen, the CampyGen also requires no catalyst and no water activation steps. No dangerous gas is produced. The final atmosphere is reduced to 6% oxygen, and supplemented with 14% carbon dioxide which is ideal for growth of Campylobacter and other microaerophilic organisms. Sachet, bags and clips are sold separately.
 : Pouches and clips sold separately.
Documents
Spécifications
  • Désignation:
    CampyGen compact
  • Taille:
    20 sachets