Dropping Bottles
Amber, 60 ml
Catalog # 470217-654
Supplier: Wards
Fournisseur #: CH0187F
Jump to:
Product Details & Documents
Dropping bottles are available in clear or amber formats.
- Screw cap included
- Dropper included
Bottles are made of soda glass.
Spécifications
- Désignation:Amber, 60 ml