Dropping Bottles | Avantor
Dropping Bottles
Amber, 60 ml
Catalog # 470217-654
Supplier:   Wards
Fournisseur #:  CH0187F
Dropping Bottles
Catalog # 470217-654
Supplier:  Wards
Dropping Bottles
Dropping Bottles
Amber, 60 ml
Price
$5.60
Log in pour la disponibilité
Each
Product Details & Documents
Dropping bottles are available in clear or amber formats.
  • Screw cap included
  • Dropper included
Bottles are made of soda glass.
Spécifications
  • Désignation:
    Amber, 60 ml