Dropping Bottles
Clear, 100 ml
Catalog # 470217-650
Supplier #: CH0187C
Jump to:
Discontinued
Product Details & Documents
Dropping bottles are available in clear or amber formats.
- Screw cap included
- Dropper included
Bottles are made of soda glass.
Specifications
- Désignation:Clear, 100 ml