Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
GBS Detect/Strep Reagents

GBS Detect/Strep Reagents

Granada Medium, for Hemolytic Group B Strep, 15×100 mm plate, Pack of 10
Catalog # CA10001-426
Supplier #:  G123
GBS Detect/Strep Reagents
Catalog # CA10001-426
CAS Number:  
GBS Detect/Strep Reagents
GBS Detect/Strep Reagents

Granada Medium, for Hemolytic Group B Strep, 15×100 mm plate, Pack of 10

CAS Number:  
Price
$73.60
Log in for availability
Pack of 10
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

GBS Detect is used for the isolation and differentiation by enhanced hemolysis of gamma hemolytic Group B Streptococcus.

  • Grouping reagents for the rapid latex agglutination test
  • Lancefield grouping includes A,B,C,D,F,G latex, controls, extraction reagents, cards, and mixing sticks
  • StrepQuick™ to aid in the identification of gram positive cocci

GBS Detect™, for non-hemolytic Group B Strep. Granada Medium for hemolytic Group B Strep. Grouping Reagents A, B, C, and G, for the rapid latex agglutination test. PYR Test Kit with chromogenic developer on paper disks. Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), especially for throat cultures. Selective Strep Agar (COBA Medium), for the selective isolation of all species of Strep. ci. StrepQuick™ performs a PYR, LAP, and Esculin test to aid in the identification of gram positive coc.

Specifications

Specifications

  • Désignation:
    Granada Medium, for Hemolytic Group B Strep
  • Taille:
    Pack of 10
  • Format:
    15×100 mm plate

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.