Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape), 11 mm, Black, Snap cap
10803-618
: 9502C-E-30KX-M
- Taille du bouchon:11 mm
- Cap Type:Snap cap
- Septum:Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape)
- Matière:PE
- Cap Color:Black