Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape), 11 mm, Black, Snap cap
Catalog # 10803-616
Supplier: MicroSolv Technology Corp
Fournisseur #: 9502C-E-30KX
Jump to:
Product Details & Documents
Spécifications
- Taille du bouchon:11 mm
- Cap Type:Snap cap
- Septum:Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit (X Shape)
- Matière:PE
- Cap Color:Black