Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit, 11 mm, Black, Snap cap
Catalog # 10803-614
Supplier: MicroSolv Technology Corp
: 9502C-E-30K-M
Jump to:
Product Details & Documents
Spécifications
- Taille du bouchon:11 mm
- Cap Type:Snap cap
- Septum:Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit
- Matière:PE
- Cap Color:Black