Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE, 11 mm, Blue, Snap cap
10803-562
: 9502C-E-10B-M
For use with 12 x 32 mm autosampler vials.
- Taille du bouchon:11 mm
- Cap Type:Snap cap
- Septum:Soft-Guard, White Silicone Rubber / Red PTFE
- Matière:PE
- Cap Color:Blue