Protocol Station, Restek
Protocol Station, Restek
Protocol Station*, Stainless steel, CGA 580 (N2, He, Ar)
Catalog # 10803-242
Supplier:   Restek
Supplier #:  21327
Protocol Station, Restek
Catalog # 10803-242
Supplier:  Restek
Protocol Station, Restek
Protocol Station, Restek
Protocol Station*, Stainless steel, CGA 580 (N2, He, Ar)
Price
$1,540.80
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
  • Fabricated in stainless steel or chrome-plated brass
  • Provides ease of use for high-purity gas regulators
Wall mounting provides ease of use, prevents gas regulator damage, and improves safety. Either chrome-plated brass or 316 stainless steel option is complete with a 3-foot, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.
Ordering information: Pressure regulator not included. Order separately.
Specifications
  • Désignation:
    Protocol Station*
  • Matière:
    Stainless steel
  • Includes:
    CGA 580 (N2, He, Ar)