Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SDS Binders | Avantor

SDS Binders

SDS Binder with 36" Metal Chain, Holds up to 600 pages, 3"
Catalog # 470165-244
Fournisseur #:  SDSB3
SDS Binders
Catalog # 470165-244
Supplier:  
SDS Binders
SDS Binders

SDS Binder with 36" Metal Chain, Holds up to 600 pages, 3"

Discontinued
Product Details & Documents

SDS Binders keep essential SDS information organized and safe.

  • Binders are clearly labeled
  • Sheet protectors available
  • SDS info safeguarded

The clearly labeled binders make it easy to store and access information.

They are available with and without sheet protectors. It is a sturdy plastic binder protects sheets from use and abuse.

Spécifications
  • Sheet Capacity:
    Holds up to 600 pages
  • Désignation:
    SDS Binder with 36" Metal Chain
  • Taille:
    3"