Human Recombinant FGF-basic (from E. coli), PeproTech
500 μg, US
10771-952
: 100-18C-500UG
FGF-basic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-basic is a non-glycosylated, heparin-binding growth factor that is expressed in the brain, pituitary, kidney, retina, bone, testis, adrenal gland, liver, monocytes, epithelial cells and endothelial cells. FGF-basic signals through FGFR 1b, 1c, 2c, 3c and 4. Recombinant Human FGF-basic is a 16.4 kDa protein consisting of 146 amino acid residues.
: For research use only. Not for use in diagnostic or therapeutic procedures.
- Taille:500 μg
- Origin:US
- Protease-free:Y
- Form:Lyophilized
- Protein/peptide name:FGF-basic
- Protein synonyms:Prostatropin|HBGF-2|Fibroblast Growth Factor-basic|FGF-2
- Protein Function:Cytokines and Growth Factors
- Purity:0.98
- Endotoxin level:Low
- Sequence:PALPEDGGSG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH IKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTD ECFFFERLES NNYNTYRSRK YTSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- Source:E. coli
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Molecular weight:16.4 kDa
- Shipping temperature:Ambient
- Cat. No.:10771-952
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protein/peptide type:Recombinant
- Endotoxin content:>0.1 ng/μg
- Storage conditions:Store at –20 °C prior to reconstitution.
- Gene ID:2247 P09038
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Conjugation:Unconjugated
- Species:Human



