Human Recombinant FGF-basic (from E. coli), PeproTech
250 μg, US
Catalog # 10771-950
Supplier: Peprotech
Fournisseur #: 100-18C-250UG
Human Recombinant FGF-basic (from <i>E. coli</i>), PeproTech
Catalog # 10771-950
Supplier: Peprotech
CAS Number:
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
FGF-basic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-basic is a non-glycosylated, heparin-binding growth factor that is expressed in the brain, pituitary, kidney, retina, bone, testis, adrenal gland, liver, monocytes, epithelial cells and endothelial cells. FGF-basic signals through FGFR 1b, 1c, 2c, 3c and 4. Recombinant Human FGF-basic is a 16.4 kDa protein consisting of 146 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Spécifications
Spécifications
- Taille:250 μg
- Origin:US
- Protease-free:Y
- Form:Lyophilized
- Protein/peptide name:FGF-basic
- Protein synonyms:Prostatropin|HBGF-2|Fibroblast Growth Factor-basic|FGF-2
- Protein Function:Cytokines and Growth Factors
- Purity:0.98
- Endotoxin level:Low
- Sequence:PALPEDGGSG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH IKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTD ECFFFERLES NNYNTYRSRK YTSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- Source:E. coli
- Biological activity:Biological Activity tests can vary. Please refer to the Product CoA for information.
- Molecular weight:16.4 kDa
- Shipping temperature:Ambient
- Cat. No.:10771-950
- Reconstitution instructions:Reconstitution instructions can vary. Please refer to the product CoA for info.
- Protein/peptide type:Recombinant
- Endotoxin content:>0.1 ng/μg
- Storage conditions:Store at –20 °C prior to reconstitution.
- Gene ID:2247 P09038
- Formulation:Formulation can vary. Please refer to product CoA for information.
- Conjugation:Unconjugated
- Species:Human



