Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant FGF-basic (from E. coli), PeproTech | Avantor

Human Recombinant FGF-basic (from E. coli), PeproTech

10 μg, US
Catalog # 10771-944
Supplier:   Peprotech
Fournisseur #:  100-18C-10UG
Human Recombinant FGF-basic (from <i>E. coli</i>), PeproTech
Catalog # 10771-944
Supplier:  Peprotech
CAS Number:  
Human Recombinant FGF-basic (from E. coli), PeproTech

10 μg, US

CAS Number:  
Price
$256.23
Log in pour la disponibilité
Each (10µG)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

FGF-basic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-basic is a non-glycosylated, heparin-binding growth factor that is expressed in the brain, pituitary, kidney, retina, bone, testis, adrenal gland, liver, monocytes, epithelial cells and endothelial cells. FGF-basic signals through FGFR 1b, 1c, 2c, 3c and 4. Recombinant Human FGF-basic is a 16.4 kDa protein consisting of 146 amino acid residues.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Spécifications

Spécifications

  • Taille:
    10 μg
  • Origin:
    US
  • Protease-free:
    Y
  • Form:
    Lyophilized
  • Purity:
    0.98
  • Source:
    E. coli
  • Molecular weight:
    16.4 kDa
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    >0.1 ng/μg
  • Gene ID:
    2247 P09038
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Species:
    Human
  • Protein/peptide name:
    FGF-basic
  • Protein synonyms:
    Prostatropin|HBGF-2|Fibroblast Growth Factor-basic|FGF-2
  • Protein Function:
    Cytokines and Growth Factors
  • Endotoxin level:
    Low
  • Sequence:
    PALPEDGGSG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH IKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTD ECFFFERLES NNYNTYRSRK YTSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Shipping temperature:
    Ambient
  • Cat. No.:
    10771-944
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Conjugation:
    Unconjugated

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.