Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Cleanroom Papers | Avantor

VWR® Cleanroom Papers

Latex-free paper, Blue, 11×17", 22 lbs.
Catalog # 10769-750
Supplier:   VWR International
VWR® Cleanroom Papers
Catalog # 10769-750
Supplier:  VWR International
VWR® Cleanroom Papers
VWR® Cleanroom Papers

Latex-free paper, Blue, 11×17", 22 lbs.

Price
$646.62
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1,250
Product Details & Documents

The cleanroom paper is ideal for documentation in critical environments, where very low particle and ESD generation is important.

  • Low particle generation

Cleanroom paper is provided in latex or latex-free variants in a variety of colors.

Spécifications
  • Color:
    Blue
  • Désignation:
    Latex-free paper
  • Dimensions:
    11×17"
  • Weight:
    22 lbs.