Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SpillStopper Work Surfaces, Labconco | Avantor

SpillStopper Work Surfaces, Labconco

Work Surface-LR,RS, 36", 36", Left rear and right side
Catalog # 10769-472
Supplier:   Labconco
Fournisseur #:  9501312
SpillStopper Work Surfaces, Labconco
Catalog # 10769-472
Supplier:  Labconco
SpillStopper Work Surfaces, Labconco
SpillStopper Work Surfaces, Labconco

Work Surface-LR,RS, 36", 36", Left rear and right side

Price
$3,320.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

SpillStopper Work Surfaces are molded from a special formulation of corrosion-resistant epoxy resins

Dished and contoured to conform to the interior liner of Protector Hoods. Front edge has a large radius to aerodynamically direct airflow into the hood. Pre-drilled 1.5" (5.8 cm) diameter holes for venting and 2.5" (6.4 cm) diameter holes for plumbing pass-through.

Ordering information: May be ordered with a pre-cut 15.2 x 7.6 cm (6 x 3") oval cupsink cutout. Cupsink is sold separately. SpillStopper Work Surfaces provide support for Protector Premier, XStream, XL, Echo and Airo Laboratory Hoods.

Spécifications
  • Désignation:
    Work Surface-LR,RS
  • Profondeur:
    36"
  • Width:
    36"
  • Cupsink Cutout:
    Left rear and right side
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.