Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
SpillStopper Work Surfaces, Labconco | Avantor

SpillStopper Work Surfaces, Labconco

Work Surface-LS,RS, 36", 96", Left side and right side
Catalog # 10769-458
Supplier:   Labconco
Fournisseur #:  9501822
SpillStopper Work Surfaces, Labconco
Catalog # 10769-458
Supplier:  Labconco
SpillStopper Work Surfaces, Labconco
SpillStopper Work Surfaces, Labconco

Work Surface-LS,RS, 36", 96", Left side and right side

Price
$6,570.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

SpillStopper Work Surfaces are molded from a special formulation of corrosion-resistant epoxy resins

Dished and contoured to conform to the interior liner of Protector Hoods. Front edge has a large radius to aerodynamically direct airflow into the hood. Pre-drilled 1.5" (5.8 cm) diameter holes for venting and 2.5" (6.4 cm) diameter holes for plumbing pass-through.

 : May be ordered with a pre-cut 15.2 x 7.6 cm (6 x 3") oval cupsink cutout. Cupsink is sold separately. SpillStopper Work Surfaces provide support for Protector Premier, XStream, XL, Echo and Airo Laboratory Hoods.

Spécifications
  • Désignation:
    Work Surface-LS,RS
  • Profondeur:
    36"
  • Width:
    96"
  • Cupsink Cutout:
    Left side and right side
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.