SpillStopper Work Surfaces, Labconco
Work Surface-LR,RS, 36", 96", Left rear and right side
  10769-414
 :   Labconco
 :  9501812
SpillStopper Work Surfaces, Labconco
  10769-414
 :  Labconco
SpillStopper Work Surfaces, Labconco
SpillStopper Work Surfaces, Labconco
Work Surface-LR,RS, 36", 96", Left rear and right side
6 570,00 $
Each
SpillStopper Work Surfaces are molded from a special formulation of corrosion-resistant epoxy resins
Dished and contoured to conform to the interior liner of Protector Hoods. Front edge has a large radius to aerodynamically direct airflow into the hood. Pre-drilled 1.5" (5.8 cm) diameter holes for venting and 2.5" (6.4 cm) diameter holes for plumbing pass-through.
 : May be ordered with a pre-cut 15.2 x 7.6 cm (6 x 3") oval cupsink cutout. Cupsink is sold separately. SpillStopper Work Surfaces provide support for Protector Premier, XStream, XL, Echo and Airo Laboratory Hoods.
  • Désignation:
    Work Surface-LR,RS
  • Profondeur:
    36"
  • Width:
    96"
  • Cupsink Cutout:
    Left rear and right side