Variocool Chillers, LAUDA-Brinkmann
Variocool Chillers, LAUDA-Brinkmann
Catalog # 10768-430
Variocool Chillers, LAUDA-Brinkmann
Catalog # 10768-430
Supplier:  
Variocool Chillers, LAUDA-Brinkmann
Discontinued
Product Details & Documents
LAUDA Variocool chillers offer a broad performance range for demanding temperature control tasks.
  • Color screen controller is both intuitive and informative
  • USB interface and alarm contact are standard features built into each unit
  • Option of adding additional communication interfaces for increased flexibility
LAUDA Variocool chillers were designed with a modular concept and offer a broad range of options. High powered pumps and heaters allow for a truly custom design to match your application's needs.
Certifications: CE compliant.
Ordering information: Chiller is supplied with USB and alarm contact standard. All units listed below are in 60Hz frequencies. Variant units in 50Hz configurations are also available upon request. Optional heaters and pumps are sold separately, except chiller model VC 5000 (L000756), which includes a heater (model LWZ 3096).