Variable Volume Pipettes, Hach
Variable Volume Pipette Set with Tips, 1× each 0.2 - 1.0 ml and 1.0 - 5.0 ml
Catalog # CA89501-142
Jump to:
Discontinued
Product Details & Documents
Hach's variable volume single-channel, air-displacement pipettes feature robust design, comfortable ergonomics, slim-line tip ejection, and single-handed operation.
Outstanding accuracy with adjustments in 50 L increments (0.05 ml).
: Supplied with calibration report and maintenance tool.
Spécifications
- Volume:1× each 0.2 - 1.0 ml and 1.0 - 5.0 ml
- Désignation:Variable Volume Pipette Set with Tips