pH Buffer Solutions, Hach
±0.02, 25 °C, 4.01
Catalog # CA10199-498
pH Buffer Solutions, Hach
Catalog # CA10199-498
Supplier:  
CAS Number:  
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach
±0.02, 25 °C, 4.01
CAS Number:  
Discontinued
Product Details & Documents
For pH meter standardization
Available colorless, or color-coded for easy identification.
Documents
Spécifications
Spécifications
  • Reference Temperature:
    25 °C
  • pH:
    4.01
  • pH Accuracy:
    ±0.02