ASTM Class 7 Hooked Weights
0.2 N
Catalog # 10754-376
Supplier: Troemner
Supplier #: 30390783
Jump to:
Product Details & Documents
ASTM hooked weights for weighing operations in physical and chemical classrooms, such as force measuring.
- 303 stainless steel
- ASTM Class 7
All surfaces are satin finish and meet or exceed ANSI/ASTM E617 tolerance and design restrictions.
Specifications
- Weight:0.2 N