ASTM Class 7 Hooked Weights | Avantor
ASTM Class 7 Hooked Weights
1 N
Catalog # 10754-372
Supplier:   Troemner
Fournisseur #:  30390781
ASTM Class 7 Hooked Weights
Catalog # 10754-372
Supplier:  Troemner
ASTM Class 7 Hooked Weights
ASTM Class 7 Hooked Weights
1 N
Price
$66.71
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
ASTM hooked weights for weighing operations in physical and chemical classrooms, such as force measuring.
  • 303 stainless steel
  • ASTM Class 7
All surfaces are satin finish and meet or exceed ANSI/ASTM E617 tolerance and design restrictions.
Spécifications
  • Weight:
    1 N