Sterikon® Plus Bioindicators, MilliporeSigma | Avantor
Sterikon® Plus Bioindicators, MilliporeSigma
100 Ampules
Catalog # CA10752-390
Supplier:   MilliporeSigma
Fournisseur #:  1.10274.0002
Sterikon® Plus Bioindicators, MilliporeSigma
Catalog # CA10752-390
Supplier:  MilliporeSigma
Sterikon® Plus Bioindicators, MilliporeSigma
Sterikon® Plus Bioindicators, MilliporeSigma
100 Ampules
Price
$1,102.71
Log in pour la disponibilité
Each
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
A rapid and easy-to-use method for verifying steam sterilization.
Indicators consist of an ampule containing nutrient broth, sugar, a pH indicator, and 2mL of spores from an apathogenic organism: Bacillus stearothermophilus ATCC 7953
Use to monitor the effectiveness of steam sterilization at 121°C (250°F) for 15 minutes, as well as to confirm that the prescribed temperature is reached throughout the chamber and remains constant for the entire 15 minutes. Autoclaves with capacities of up to 250 L require at least two ampules, while larger ones require at least six.
Note: Not intended for monitoring sterilization of medical devices — for laboratory use only. Avoid use of the ampules at temperatures exceeding 125°C (257°F) to prevent damage to the bioindicator, resulting in color changes without increased turbidity.
Simply place ampules in the autoclave along with the batch to be sterilized, and incubate afterwards. A color change of the ampule contents clearly indicates whether sterilization was successful.
Certifications: Sterikon Plus meets USP XXIV guidelines for monitoring the effectiveness of steam sterilization.
Documents
Spécifications
  • Taille:
    100 Ampules
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.