Saline Solutions, NERL Diagnostics
10 L (2.64 gal.)
Supplier: Thermo Fisher Scientific
Fournisseur #: 850510L
Saline Solutions, NERL Diagnostics
Catalog # CA48212-271L
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Blood bank saline pH 7.0–7.2 is a phosphate buffered saline solution manufactured to match the pH of human blood
Manufactured to meet NCCLS standard ASI/BB-1. Certificates of analysis are available.
Note: Not for infusion, irrigation, or injection.
This preservative-free sodium chloride solution is for use in serological (blood bank) testing or for general laboratory use.
Documents
Certificates
Recherche par numéro de lotSpécifications
Spécifications
- Volume:10 L (2.64 gal.)



