Student gratings
Student gratings
Student gratings, 100 lines per mm
Catalog # 470158-408
Supplier #:  DFG100
Student gratings
Catalog # 470158-408
Student gratings
Student gratings
Student gratings, 100 lines per mm
Price
$85.65
Log in pour la disponibilité
Each
Product Details & Documents
Quality gratings perfect for your Optics Needs
  • Viewing area measures approx. 45x30mm
Quality transmission gratings protected by two glass plates, 60x45mm.
Spécifications
  • Désignation:
    Student gratings, 100 lines per mm