Student gratings
Student gratings, 100 lines per mm
Catalog # 470158-408
Supplier: United Scientific Supplies
Supplier #: DFG100
Jump to:
Product Details & Documents
Quality gratings perfect for your Optics Needs
- Viewing area measures approx. 45x30mm
Quality transmission gratings protected by two glass plates, 60x45mm.
Spécifications
- Désignation:Student gratings, 100 lines per mm