Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Precision Air Table

Precision Air Table

Precision Air Table
Catalog # 470157-648
Supplier:   Wards
Supplier #:  AIRTBL
Precision Air Table
Catalog # 470157-648
Supplier:  Wards
Precision Air Table
Precision Air Table

Precision Air Table

Price
$1,415.20
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

The air table guides the study of a multitude of concepts.

  • Topics Include Elastic and Inelastic Collisions, Linear and Angular Momentum, and More
  • Air Table Length: 700 mm
  • Width: 500 mm
  • Adjustable Feet

These concepts include elastic and inelastic collisions, linear and angular momentum, conservation of momentum, moment of inertia, centripetal force, and simple and complex harmonic motion.

The air table is 500 x 700 mm with adjustable feet. It requires an air blower.

Specifications
  • Désignation:
    Precision Air Table
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.