VWR® Round-Bottom Boiling Flasks | Avantor
VWR® Round-Bottom Boiling Flasks
2000 ml, 24/40
Catalog # 10536-674
Supplier:   VWR International
VWR® Round-Bottom Boiling Flasks
Catalog # 10536-674
Supplier:  VWR International
VWR® Round-Bottom Boiling Flasks
VWR® Round-Bottom Boiling Flasks
2000 ml, 24/40
Price
$172.27
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 1 Case of 6
Product Details & Documents
Durable boiling flasks have good temperature and chemical resistance.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403 Type 1. The flasks feature uniform wall thickness, which makes them ideal for heating.
Documents
Spécifications
  • Capacité:
    2000 ml
  • Neck Size:
    24/40