VWR® Round-Bottom Boiling Flasks
200 ml, 24/40
Supplier: VWR International
Jump to:
Product Details & Documents
Durable boiling flasks have good temperature and chemical resistance.
- Good temperature resistance
- Chemically resistant
- Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403 Type 1. The flasks feature uniform wall thickness, which makes them ideal for heating.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Capacité:200 ml
- Neck Size:24/40