VWR® Flat-Bottom Boiling Flasks
VWR® Flat-Bottom Boiling Flasks
300 ml, 24/40
  10536-638
VWR® Flat-Bottom Boiling Flasks
  10536-638
VWR® Flat-Bottom Boiling Flasks
VWR® Flat-Bottom Boiling Flasks
300 ml, 24/40
$133.17
Pack of 2 Case of 12
The flasks come with uniform wall thickness which makes these flasks ideal for heating.
  • Borosilicate glass fabrication
  • Good chemical resistance
Durable product has good temperature and chemical resistance.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403, Type 1 Class 4.
Documents
  • Capacité:
    300 ml
  • Neck Size:
    24/40