Duramark® Lock Boxes
Metal lock box with combination lock, 35.5×26.7×17.8 cm (14×10½×7”)
Catalog # 10497-138
Supplier: Therapak, LLC
Fournisseur #: 36619
Jump to:
Product Details & Documents
Metal lock boxes are constructed of 24 gauge cold-rolled steel, painted with a powder coat finish, and insulated with foam
Plastic lock boxes are made of high impact polystrene and feature a viewing window on the door. Both boxes feature a hinged door that opens downward for convenient access and use as a tray. Two matching keys are supplied. The metal lock box is also available with a combination lock. Over- or under-door straps allow the boxes to be secured outside of a locked door for easy courier access.
Spécifications
- Désignation:Metal lock box with combination lock
- Dimensions:35.5×26.7×17.8 cm (14×10½×7”)
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Under-Door Strap for Metal or Plastic Box
Informations sur le produit
- Catalog #AvailabilityPrice
- Over-Door Strap for Metal or Plastic Box
Informations sur le produit
- Catalog #AvailabilityPrice
- Extra Long Over-Door Strap for Metal or Plastic BoxExtra Long Over-Door Strap for Metal or Plastic Box10497-106Log in pour la disponibilitéEach - $188.06
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement Key for Metal or Plastic Box



