Basic Upright Frame
Auxiliary top bracket, left
Catalog # 10496-886
Supplier: Treston
: 92641001
Jump to:
Product Details & Documents
Basic™ upright tables are ideal for lighter use, when some additional workspace is needed.
- Bench top attached to the frame, can be equipped with any Treston accessories
- May also be used without a worksurface as accessory support positioned behind conveyors or machines
The slotted upright frame allows for the use of different accessories.
: Components sold separately. Installation is required. Assembly videos available.
Spécifications
- Désignation:Auxiliary top bracket, left