Basic Upright Frame | Avantor
Basic Upright Frame
Catalog # 10496-634
Basic Upright Frame
Catalog # 10496-634
Supplier:  
Basic Upright Frame
Basic Upright Frame
Discontinued
Product Details & Documents
Basic™ upright tables are ideal for lighter use, when some additional workspace is needed.
  • Bench top attached to the frame, can be equipped with any Treston accessories
  • May also be used without a worksurface as accessory support positioned behind conveyors or machines
The slotted upright frame allows for the use of different accessories.
 : Components sold separately. Installation is required. Assembly videos available.