Cables, Huber
Control cable, RS232 (9 <> 9 pin), 3 m (10 ft.)
Catalog # 10221-334
Jump to:
Discontinued
Product Details & Documents
Spécifications
- Désignation:Control cable, RS232 (9 <> 9 pin)
- Longueur:3 m (10 ft.)