Flow Indicators, Bürkle | Avantor
Flow Indicators, Bürkle
CheMobil, Black, PMP, 6 - 11 mm
Catalog # 10217-294
Supplier:   BURKLE INC
Fournisseur #:  8720-2003
Flow Indicators, Bürkle
Catalog # 10217-294
Supplier:  BURKLE INC
Flow Indicators, Bürkle
Flow Indicators, Bürkle
CheMobil, Black, PMP, 6 - 11 mm
Price
$51.95
Log in pour la disponibilité
Each
Product Details & Documents
These paddle wheel flow indicators give a clear visualisation of the flow rates of liquids or gas in tubing
LiquiMobil flow indicator made of SAN is mainly suitable for cooling systems and helps to reduce water consumption considerably because the cooling water fl ow can be adjusted better. Not suitable for aggressive substances. Maximum operating temperature LiquiMobil SAN is +30 °C.
CheMobil PMP shows reasonable chemical resistance, which makes it possible to use with a great number of fluids. CheMobil is not however appropriate for strong acids, bases and solvents. CheMobil is steam sterilisable. Maximum operating temperature +60 °C.
The flow indicator can operate in either direction of flow and can be mounted at any angle. The paddle wheel impeller starts to rotate at about 150 ml/min. Colour coding - red, black or blue
Spécifications
  • Type:
    CheMobil
  • Color:
    Black
  • Matière:
    PMP
  • Tube Diameter:
    6 - 11 mm